Assay Details
| Gene Symbol: LOC4342782 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789125.2 | XP_015644611.1 | 2 - 3 | 892 | 82 | |
| GenBank mRNA | AY324148.1 | - | 2 - 3 | 874 | 82 | |
| AY327038.1 | - | 3 - 4 | 804 | 82 | ||
More Information
Set Membership: |
|

LOC4342782
Genomic Map