Assay Details
| Gene Symbol: LOC107276899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789920.2 | XP_015645406.1 | 8 - 9 | 1681 | 105 | |
| XM_015789921.2 | XP_015645407.1 | 8 - 9 | 1689 | 105 | ||
| XM_015789922.2 | XP_015645408.1 | 8 - 9 | 1689 | 105 | ||
More Information
Set Membership: |
|

LOC107276899
Genomic Map