Assay Details
| Gene Symbol: LOC4343080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789420.2 | XP_015644906.1 | 10 - 11 | 1235 | 102 | |
| GenBank mRNA | AK060367.1 | - | 2 - 3 | 109 | 102 | |
| AK103253.1 | - | 10 - 11 | 1215 | 102 | ||
More Information
Set Membership: |
|

LOC4343080
Genomic Map