Assay Details
| Gene Symbol: LOC9269328 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790705.2 | XP_015646191.1 | 12 - 13 | 1076 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC9269328| Gene Symbol: LOC9269328 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790705.2 | XP_015646191.1 | 12 - 13 | 1076 | 89 | |
Set Membership: |
|