Assay Details
| Gene Symbol: LOC4343617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789328.2 | XP_015644814.1 | 3 - 4 | 525 | 72 | |
| GenBank mRNA | AK061125.1 | - | 3 - 4 | 316 | 72 | |
| AK102110.1 | - | 3 - 4 | 531 | 72 | ||
More Information
Set Membership: |
|

LOC4343617
Genomic Map