Assay Details
| Gene Symbol: LOC4343617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789328.2 | XP_015644814.1 | 8 - 9 | 891 | 110 | |
| GenBank mRNA | AK061125.1 | - | 8 - 9 | 682 | 110 | |
| AK102110.1 | - | 8 - 9 | 897 | 110 | ||
More Information
Set Membership: |
|

LOC4343617
Genomic Map