Assay Details
Gene Symbol: LOC9271563 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015789244.2 | XP_015644730.1 | 4 - 5 | 637 | 77 | |
GenBank mRNA | AK107534.1 | - | 3 - 4 | 670 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
