Assay Details
Gene Symbol: LOC9271563 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015789244.2 | XP_015644730.1 | 5 - 6 | 881 | 84 | |
GenBank mRNA | AK107534.1 | - | 4 - 5 | 914 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
