Assay Details
| Gene Symbol: LOC4343985 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790209.2 | XP_015645695.1 | 2 - 3 | 307 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4343985| Gene Symbol: LOC4343985 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790209.2 | XP_015645695.1 | 2 - 3 | 307 | 101 | |
Set Membership: |
|