Assay Details
| Gene Symbol: LOC4344175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791202.2 | XP_015646688.1 | 26 - 27 | 4388 | 82 | |
| GenBank mRNA | AK063537.1 | - | 2 - 3 | 197 | 82 | |
| AK067331.1 | - | 8 - 9 | 1179 | 82 | ||
More Information
Set Membership: |
|

LOC4344175
Genomic Map