Assay Details
| Gene Symbol: LOC4344219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790708.2 | XP_015646194.1 | 2 - 3 | 808 | 145 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4344219| Gene Symbol: LOC4344219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790708.2 | XP_015646194.1 | 2 - 3 | 808 | 145 | |
Set Membership: |
|