Assay Details
| Gene Symbol: LOC4344231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791332.1 | XP_015646818.1 | 13 - 14 | 1007 | 80 | |
| GenBank mRNA | AK072249.1 | - | 13 - 14 | 1014 | 80 | |
| AK104085.1 | - | 7 - 8 | 424 | 80 | ||
More Information
Set Membership: |
|

LOC4344231
Genomic Map