Assay Details
| Gene Symbol: LOC4344231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791332.1 | XP_015646818.1 | 19 - 20 | 1428 | 95 | |
| GenBank mRNA | AK072249.1 | - | 19 - 20 | 1435 | 95 | |
| AK104085.1 | - | 13 - 14 | 845 | 95 | ||
More Information
Set Membership: |
|

LOC4344231
Genomic Map