Assay Details
| Gene Symbol: LOC4344241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789588.2 | XP_015645074.1 | 1 - 2 | 666 | 75 | |
| GenBank mRNA | AK060349.1 | - | 1 - 2 | 635 | 75 | |
| AK102921.1 | - | 1 - 2 | 637 | 75 | ||
More Information
Set Membership: |
|

LOC4344241
Genomic Map