Assay Details
| Gene Symbol: LOC4344342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791262.2 | XP_015646748.1 | 8 - 9 | 851 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4344342| Gene Symbol: LOC4344342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791262.2 | XP_015646748.1 | 8 - 9 | 851 | 102 | |
Set Membership: |
|