Assay Details
| Gene Symbol: LOC4344443 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792792.2 | XP_015648278.1 | 1 - 2 | 379 | 70 | |
| GenBank mRNA | AB517626.1 | - | 1 - 2 | 335 | 70 | |
| AB517627.1 | - | 1 - 2 | 335 | 70 | ||
| AK069782.1 | - | 1 - 2 | 385 | 70 | ||
| AK102613.1 | - | 1 - 2 | 335 | 70 | ||
| HQ858842.1 | - | 1 - 2 | 97 | 70 | ||
More Information
Set Membership: |
|

LOC4344443
Genomic Map