Assay Details
| Gene Symbol: LOC4344443 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792792.2 | XP_015648278.1 | 5 - 6 | 1041 | 96 | |
| GenBank mRNA | AB517626.1 | - | 5 - 6 | 997 | 96 | |
| AB517627.1 | - | 5 - 6 | 997 | 96 | ||
| AK069782.1 | - | 5 - 6 | 1047 | 96 | ||
| AK102613.1 | - | 5 - 6 | 997 | 96 | ||
| HQ858842.1 | - | 5 - 6 | 759 | 96 | ||
More Information
Set Membership: |
|

LOC4344443
Genomic Map