Assay Details
| Gene Symbol: LOC4344482 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795342.2 | XP_015650828.1 | 14 - 15 | 2260 | 86 | |
| XM_015795343.2 | XP_015650829.1 | 14 - 15 | 2261 | 86 | ||
| GenBank mRNA | AK066813.1 | - | 14 - 15 | 2257 | 86 | |
| AK099475.1 | - | 14 - 15 | 2250 | 86 | ||
More Information
Set Membership: |
|

LOC4344482
Genomic Map