Assay Details
| Gene Symbol: LOC4344482 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795342.2 | XP_015650828.1 | 21 - 22 | 2952 | 147 | |
| GenBank mRNA | AK066813.1 | - | 21 - 22 | 2949 | 147 | |
| AK099475.1 | - | 21 - 22 | 2942 | 147 | ||
More Information
Set Membership: |
|

LOC4344482
Genomic Map