Assay Details
| Gene Symbol: LOC4344482 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795342.2 | XP_015650828.1 | 2 - 3 | 497 | 82 | |
| XM_015795343.2 | XP_015650829.1 | 2 - 3 | 498 | 82 | ||
| GenBank mRNA | AK066813.1 | - | 2 - 3 | 494 | 82 | |
| AK099475.1 | - | 2 - 3 | 487 | 82 | ||
More Information
Set Membership: |
|

LOC4344482
Genomic Map