Assay Details
Gene Symbol: LOC4344647 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792962.2 | XP_015648448.1 | 9 - 10 | 1162 | 72 | |
GenBank mRNA | AK106434.1 | - | 9 - 10 | 1145 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
