Assay Details
| Gene Symbol: LOC4344688 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792717.2 | XP_015648203.1 | 7 - 8 | 871 | 64 | |
| GenBank mRNA | AK060859.1 | - | 7 - 8 | 871 | 64 | |
| AK099613.1 | - | 7 - 8 | 731 | 64 | ||
More Information
Set Membership: |
|

LOC4344688
Genomic Map