Assay Details
| Gene Symbol: LOC4344728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB638269.1 | - | 13 - 14 | 1529 | 85 | |
| AK111830.1 | - | 13 - 14 | 2086 | 85 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4344728| Gene Symbol: LOC4344728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB638269.1 | - | 13 - 14 | 1529 | 85 | |
| AK111830.1 | - | 13 - 14 | 2086 | 85 | ||
Set Membership: |
|