Assay Details
| Gene Symbol: LOC4344728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB638269.1 | - | 18 - 19 | 2330 | 65 | |
| AK111830.1 | - | 18 - 19 | 2887 | 65 | ||
| AK111915.1 | - | 2 - 3 | 184 | 65 | ||
More Information
Set Membership: |
|

LOC4344728
Genomic Map