Assay Details
| Gene Symbol: LOC4344728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB638269.1 | - | 24 - 25 | 3200 | 69 | |
| AK111830.1 | - | 24 - 25 | 3757 | 69 | ||
| AK111915.1 | - | 8 - 9 | 1054 | 69 | ||
More Information
Set Membership: |
|

LOC4344728
Genomic Map