Assay Details
| Gene Symbol: LOC4345194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793556.2 | XP_015649042.1 | 1 - 2 | 785 | 63 | |
| GenBank mRNA | AK073881.1 | - | 1 - 2 | 674 | 63 | |
More Information
Set Membership: |
|

LOC4345194
Genomic Map