Assay Details
| Gene Symbol: LOC4345225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795516.2 | XP_015651002.1 | 2 - 3 | 460 | 97 | |
| XM_015795518.2 | XP_015651004.1 | 2 - 3 | 469 | 97 | ||
| GenBank mRNA | AK063295.1 | - | 2 - 3 | 302 | 97 | |
More Information
Set Membership: |
|

LOC4345225
Genomic Map