Assay Details
| Gene Symbol: LOC4345225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795516.2 | XP_015651002.1 | 4 - 5 | 586 | 92 | |
| XM_015795518.2 | XP_015651004.1 | 4 - 5 | 595 | 92 | ||
| GenBank mRNA | AK063295.1 | - | 4 - 5 | 428 | 92 | |
More Information
Set Membership: |
|

LOC4345225
Genomic Map