Assay Details
| Gene Symbol: LOC4345241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795318.2 | XP_015650804.1 | 6 - 7 | 763 | 96 | |
| XM_015795319.2 | XP_015650805.1 | 6 - 7 | 980 | 96 | ||
| GenBank mRNA | AK121969.1 | - | 6 - 7 | 612 | 96 | |
More Information
Set Membership: |
|

LOC4345241
Genomic Map