Assay Details
| Gene Symbol: LOC4345241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795318.2 | XP_015650804.1 | 7 - 8 | 822 | 79 | |
| XM_015795319.2 | XP_015650805.1 | 7 - 8 | 1039 | 79 | ||
| GenBank mRNA | AK121969.1 | - | 7 - 8 | 671 | 79 | |
More Information
Set Membership: |
|

LOC4345241
Genomic Map