Assay Details
| Gene Symbol: LOC4345311 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792678.1 | XP_015648164.1 | 3 - 4 | 998 | 76 | |
| GenBank mRNA | AK068483.1 | - | 3 - 4 | 1004 | 76 | |
More Information
Set Membership: |
|

LOC4345311
Genomic Map