Assay Details
| Gene Symbol: LOC4346273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792941.2 | XP_015648427.1 | 10 - 11 | 984 | 68 | |
| GenBank mRNA | AK111661.1 | - | 10 - 11 | 962 | 68 | |
| AK112044.1 | - | 10 - 11 | 961 | 68 | ||
More Information
Set Membership: |
|

LOC4346273
Genomic Map