Assay Details
Gene Symbol: LOC4346273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792941.2 | XP_015648427.1 | 1 - 2 | 150 | 126 | |
GenBank mRNA | AK111661.1 | - | 1 - 2 | 128 | 126 | |
AK112044.1 | - | 1 - 2 | 127 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
