Assay Details
| Gene Symbol: LOC4346273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792941.2 | XP_015648427.1 | 4 - 5 | 486 | 62 | |
| GenBank mRNA | AK111661.1 | - | 4 - 5 | 464 | 62 | |
| AK112044.1 | - | 4 - 5 | 463 | 62 | ||
More Information
Set Membership: |
|

LOC4346273
Genomic Map