Assay Details
| Gene Symbol: LOC4347382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755243.2 | XP_015610729.1 | 3 - 4 | 4808 | 58 | |
| XM_015755244.2 | XP_015610730.1 | 5 - 6 | 3495 | 58 | ||
| XM_015755246.2 | XP_015610732.1 | 4 - 5 | 4562 | 58 | ||
| XM_015755247.2 | XP_015610733.1 | 5 - 6 | 3485 | 58 | ||
| XM_015755248.2 | XP_015610734.1 | 3 - 4 | 3648 | 58 | ||
| GenBank mRNA | AK064218.1 | - | 1 - 2 | 1180 | 58 | |
More Information
Set Membership: |
|

LOC4347382
Genomic Map