Assay Details
| Gene Symbol: LOC4349523 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758993.2 | XP_015614479.1 | 1 - 2 | 455 | 77 | |
| GenBank mRNA | AK065306.1 | - | 1 - 2 | 414 | 77 | |
| AK104396.1 | - | 1 - 2 | 411 | 77 | ||
| AK104589.1 | - | 1 - 2 | 407 | 77 | ||
| AK104666.1 | - | 1 - 2 | 411 | 77 | ||
| AK104823.1 | - | 1 - 2 | 413 | 77 | ||
More Information
Set Membership: |
|

LOC4349523
Genomic Map