Assay Details
| Gene Symbol: LOC4323871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767021.2 | XP_015622507.1 | 4 - 5 | 702 | 72 | |
| GenBank mRNA | AK060091.1 | - | 4 - 5 | 704 | 72 | |
| AK069242.1 | - | 4 - 5 | 696 | 72 | ||
| AK100800.1 | - | 4 - 5 | 706 | 72 | ||
More Information
Set Membership: |
|

LOC4323871
Genomic Map