Assay Details
| Gene Symbol: LOC9271145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795045.2 | XP_015650531.1 | 13 - 14 | 5795 | 71 | |
| XM_015795053.2 | XP_015650539.1 | 13 - 14 | 6142 | 71 | ||
More Information
Set Membership: |
|

LOC9271145
Genomic Map