Assay Details
| Gene Symbol: LOC9271145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795045.2 | XP_015650531.1 | 14 - 15 | 5937 | 70 | |
| XM_015795053.2 | XP_015650539.1 | 14 - 15 | 6284 | 70 | ||
More Information
Set Membership: |
|

LOC9271145
Genomic Map