Assay Details
| Gene Symbol: LOC9271145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795045.2 | XP_015650531.1 | 3 - 4 | 3297 | 78 | |
| XM_015795053.2 | XP_015650539.1 | 3 - 4 | 3644 | 78 | ||
More Information
Set Membership: |
|

LOC9271145
Genomic Map