Assay Details
| Gene Symbol: LOC4324588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756364.2 | XP_015611850.1 | 2 - 3 | 1155 | 85 | |
| GenBank mRNA | AK103664.1 | - | 2 - 3 | 220 | 85 | |
| AK120836.1 | - | 2 - 3 | 879 | 85 | ||
More Information
Set Membership: |
|

LOC4324588
Genomic Map