Assay Details
| Gene Symbol: LOC4324480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766821.2 | XP_015622307.1 | 1 - 2 | 587 | 125 | |
| GenBank mRNA | AK065149.1 | - | 1 - 2 | 488 | 125 | |
| AK104357.1 | - | 1 - 2 | 487 | 125 | ||
| AK106095.1 | - | 1 - 2 | 487 | 125 | ||
More Information
Set Membership: |
|

LOC4324480
Genomic Map