Assay Details
| Gene Symbol: LOC4324480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766821.2 | XP_015622307.1 | 4 - 5 | 948 | 115 | |
| GenBank mRNA | AK065149.1 | - | 4 - 5 | 849 | 115 | |
| AK104357.1 | - | 4 - 5 | 848 | 115 | ||
| AK106095.1 | - | 4 - 5 | 848 | 115 | ||
More Information
Set Membership: |
|

LOC4324480
Genomic Map