Assay Details
| Gene Symbol: LOC4324675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761822.1 | XP_015617308.1 | 7 - 8 | 676 | 83 | |
| GenBank mRNA | AK243597.1 | - | 7 - 8 | 628 | 83 | |
More Information
Set Membership: |
|

LOC4324675
Genomic Map