Assay Details
| Gene Symbol: LOC4324149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766432.2 | XP_015621918.1 | 10 - 11 | 3253 | 80 | |
| GenBank mRNA | AK064463.1 | - | 1 - 2 | 70 | 80 | |
More Information
Set Membership: |
|

LOC4324149
Genomic Map