Assay Details
| Gene Symbol: LOC4324149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766432.2 | XP_015621918.1 | 18 - 19 | 4008 | 89 | |
| GenBank mRNA | AK064463.1 | - | 8 - 9 | 1312 | 89 | |
More Information
Set Membership: |
|

LOC4324149
Genomic Map