Assay Details
| Gene Symbol: LOC4324726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794845.2 | XP_015650331.1 | 4 - 5 | 607 | 83 | |
| GenBank mRNA | AK101326.1 | - | 4 - 5 | 475 | 83 | |
More Information
Set Membership: |
|

LOC4324726
Genomic Map