Assay Details
| Gene Symbol: LOC4325746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755778.2 | XP_015611264.1 | 5 - 6 | 870 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4325746| Gene Symbol: LOC4325746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755778.2 | XP_015611264.1 | 5 - 6 | 870 | 107 | |
Set Membership: |
|