Assay Details
| Gene Symbol: LOC4324474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793702.2 | XP_015649188.1 | 8 - 9 | 913 | 86 | |
| XM_015793709.2 | XP_015649195.1 | 9 - 10 | 825 | 86 | ||
| GenBank mRNA | AK103064.1 | - | 9 - 10 | 807 | 86 | |
More Information
Set Membership: |
|

LOC4324474
Genomic Map