Assay Details
| Gene Symbol: LOC4345814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794560.2 | XP_015650046.1 | 1 - 2 | 294 | 101 | |
| GenBank mRNA | AK073487.1 | - | 1 - 2 | 248 | 101 | |
| AK119561.1 | - | 1 - 2 | 179 | 101 | ||
| AK119761.1 | - | 1 - 2 | 180 | 101 | ||
| EU268002.1 | - | 1 - 2 | 162 | 101 | ||
More Information
Set Membership: |
|

LOC4345814
Genomic Map